siRNA Details
| Gene Symbol: Fam154b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106277.1 | NP_001099747.1 | 1, 2 | 143 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam154b| Gene Symbol: Fam154b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106277.1 | NP_001099747.1 | 1, 2 | 143 | ||