siRNA Details
| Gene Symbol: Crebzf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271791.1 | NP_001258720.1 | 1 | 764 | ||
| XM_006229688.2 | XP_006229750.1 | 1 | 1129 | |||
| XM_008759646.1 | XP_008757868.1 | 1 | 1129 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crebzf| Gene Symbol: Crebzf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271791.1 | NP_001258720.1 | 1 | 764 | ||
| XM_006229688.2 | XP_006229750.1 | 1 | 1129 | |||
| XM_008759646.1 | XP_008757868.1 | 1 | 1129 | |||