siRNA Details
| Gene Symbol: Msmb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019188.1 | NP_062061.1 | 4 | 309 | ||
| XM_008770989.1 | XP_008769211.1 | 4 | 1111 | |||
| GenBank mRNA | U65486.1 | - | 4 | 309 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Msmb| Gene Symbol: Msmb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019188.1 | NP_062061.1 | 4 | 309 | ||
| XM_008770989.1 | XP_008769211.1 | 4 | 1111 | |||
| GenBank mRNA | U65486.1 | - | 4 | 309 | ||