siRNA Details
Gene Symbol: Olr35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000121.1 | NP_001000121.1 | 1 | 508 | ||
GenBank mRNA | BC086526.1 | - | 522 | 7303669 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olr35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000121.1 | NP_001000121.1 | 1 | 508 | ||
GenBank mRNA | BC086526.1 | - | 522 | 7303669 |