siRNA Details
| Gene Symbol: Stard10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013069.1 | NP_001013087.1 | 4, 5 | 660 | ||
| XM_006229850.2 | XP_006229912.1 | 5, 6 | 783 | |||
| XM_006229851.2 | XP_006229913.1 | 4, 5 | 722 | |||
| GenBank mRNA | BC091411.1 | - | 4, 5 | 660 | 7325640 | |

Stard10
Genomic Map