siRNA Details
| Gene Symbol: Far1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011933.1 | NP_001011933.1 | 11 | 1431 | ||
| XM_006230017.2 | XP_006230079.1 | 12 | 1513 | |||
| XM_006230018.2 | XP_006230080.1 | 12 | 1438 | |||
| XM_006230020.2 | XP_006230082.1 | 12 | 1515 | |||
| GenBank mRNA | BC082015.1 | - | 11 | 1431 | 7132096 | |
| FQ221385.1 | - | 12 | 1497 | |||

Far1
Genomic Map