siRNA Details
| Gene Symbol: Olr56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000130.1 | NP_001000130.1 | 1 | 852 | ||
| Gene Symbol: LOC103690133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759842.1 | XP_008758064.1 | 1 | 534 | ||

Olr56
Genomic Map