siRNA Details
Gene Symbol: Ubqlnl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013899.1 | NP_001013921.1 | 1 | 1830 | ||
GenBank mRNA | BC083634.1 | - | 1 | 1830 | 7134647 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ubqlnl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013899.1 | NP_001013921.1 | 1 | 1830 | ||
GenBank mRNA | BC083634.1 | - | 1 | 1830 | 7134647 |