siRNA Details
Gene Symbol: Olr155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000168.1 | NP_001000168.1 | 1 | 452 | ||
XM_006229911.2 | XP_006229973.2 | 2 | 1653 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olr155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000168.1 | NP_001000168.1 | 1 | 452 | ||
XM_006229911.2 | XP_006229973.2 | 2 | 1653 |