siRNA Details
| Gene Symbol: Sigmar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030996.1 | NP_112258.1 | 2, 3 | 399 | ||
| GenBank mRNA | AF004218.1 | - | 2, 3 | 399 | ||
| AF067769.1 | - | 2, 3 | 451 | |||
| BC061978.1 | - | 2, 3 | 388 | 5623693 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sigmar1| Gene Symbol: Sigmar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030996.1 | NP_112258.1 | 2, 3 | 399 | ||
| GenBank mRNA | AF004218.1 | - | 2, 3 | 399 | ||
| AF067769.1 | - | 2, 3 | 451 | |||
| BC061978.1 | - | 2, 3 | 388 | 5623693 | ||