siRNA Details
| Gene Symbol: Olr229 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000204.1 | NP_001000204.1 | 469 | |||
| Gene Symbol: LOC100911839 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229964.2 | XP_006230026.1 | 1 | 612 | ||

Olr229
Genomic Map