siRNA Details
| Gene Symbol: Gga2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100519.1 | NP_001093989.1 | 11 | 1138 | ||
| GenBank mRNA | BC098781.1 | - | 5 | 416 | ||
| CO558907.1 | - | 533 | ||||
| Gene Symbol: LOC103694867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773816.1 | XP_008772038.1 | 10 | 1644 | ||

Gga2
Genomic Map