siRNA Details
| Gene Symbol: Fgf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022182.1 | NP_071518.1 | 1 | 363 | ||
| XM_006234903.2 | XP_006234965.1 | 2 | 1683 | |||
| GenBank mRNA | AF295300.1 | - | 1 | 331 | ||
| X56551.1 | - | 1 | 363 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fgf7| Gene Symbol: Fgf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022182.1 | NP_071518.1 | 1 | 363 | ||
| XM_006234903.2 | XP_006234965.1 | 2 | 1683 | |||
| GenBank mRNA | AF295300.1 | - | 1 | 331 | ||
| X56551.1 | - | 1 | 363 | |||