siRNA Details
| Gene Symbol: Prkacb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077645.1 | NP_001071113.1 | 9, 10 | 988 | ||
| XM_006233451.2 | XP_006233513.1 | 9, 10 | 1705 | |||
| XM_006233452.2 | XP_006233514.1 | 9, 10 | 3210 | |||
| XM_006233453.2 | XP_006233515.1 | 9, 10 | 1452 | |||
| GenBank mRNA | D10770.1 | - | 9, 10 | 988 | ||

Prkacb
Genomic Map