siRNA Details
| Gene Symbol: Bcl7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106298.1 | NP_001099768.1 | 3, 4 | 528 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bcl7c| Gene Symbol: Bcl7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106298.1 | NP_001099768.1 | 3, 4 | 528 | ||