siRNA Details
| Gene Symbol: Ahsp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106299.1 | NP_001099769.1 | 3 | 384 | ||
| GenBank mRNA | BC157822.1 | - | 3 | 308 | 8374205 | |
| FQ225822.1 | - | 3 | 305 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ahsp| Gene Symbol: Ahsp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106299.1 | NP_001099769.1 | 3 | 384 | ||
| GenBank mRNA | BC157822.1 | - | 3 | 308 | 8374205 | |
| FQ225822.1 | - | 3 | 305 | |||