siRNA Details
| Gene Symbol: Hmx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106303.1 | NP_001099773.1 | 3 | 896 | ||
| XM_008759880.1 | XP_008758102.1 | 3 | 1477 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hmx2| Gene Symbol: Hmx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106303.1 | NP_001099773.1 | 3 | 896 | ||
| XM_008759880.1 | XP_008758102.1 | 3 | 1477 | |||