siRNA Details
| Gene Symbol: Fuom | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106310.1 | NP_001099780.1 | 4 | 747 | ||
| GenBank mRNA | FQ212771.1 | - | 3 | 475 | ||
| Gene Symbol: LOC100911225 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748887.3 | XP_003748935.1 | 4 | 369 | ||

Fuom
Genomic Map