siRNA Details
| Gene Symbol: Paox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106311.1 | NP_001099781.1 | 5 | 1152 | ||
| XM_006230524.2 | XP_006230586.2 | 4 | 1127 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Paox| Gene Symbol: Paox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106311.1 | NP_001099781.1 | 5 | 1152 | ||
| XM_006230524.2 | XP_006230586.2 | 4 | 1127 | |||