siRNA Details
| Gene Symbol: Pla2g10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017176.2 | NP_058872.1 | 4 | 373 | ||
| GenBank mRNA | AF166100.1 | - | 4 | 375 | ||
| AY651029.1 | - | 4 | 369 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pla2g10| Gene Symbol: Pla2g10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017176.2 | NP_058872.1 | 4 | 373 | ||
| GenBank mRNA | AF166100.1 | - | 4 | 375 | ||
| AY651029.1 | - | 4 | 369 | |||