siRNA Details
| Gene Symbol: Rassf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106317.2 | NP_001099787.1 | 2 | 395 | ||
| XM_006230537.2 | XP_006230599.1 | 3 | 495 | |||
| XM_006230538.2 | XP_006230600.1 | 3 | 788 | |||
| XM_006230539.2 | XP_006230601.1 | 2 | 391 | |||
| GenBank mRNA | BC158743.1 | - | 2 | 395 | 9100666 | |

Rassf7
Genomic Map