siRNA Details
| Gene Symbol: Tef | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019194.2 | NP_062067.2 | 1 | 202 | ||
| GenBank mRNA | BC101895.1 | - | 1 | 202 | 7932583 | |
| S58745.1 | - | 1 | 25 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tef| Gene Symbol: Tef | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019194.2 | NP_062067.2 | 1 | 202 | ||
| GenBank mRNA | BC101895.1 | - | 1 | 202 | 7932583 | |
| S58745.1 | - | 1 | 25 | |||