siRNA Details
| Gene Symbol: Mrgprd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001506.1 | NP_001001506.1 | 1 | 668 | ||
| XM_006230725.2 | XP_006230787.1 | 2 | 1587 | |||
| GenBank mRNA | AB154411.1 | - | 1 | 668 | ||
More Information
Set Membership: |
|

Mrgprd
Genomic Map