siRNA Details
| Gene Symbol: Ndufs8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106322.2 | NP_001099792.1 | 5 | 379 | ||
| XM_006230731.1 | XP_006230793.1 | 5 | 590 | |||
| GenBank mRNA | BC158791.1 | - | 5 | 379 | 7311183 | |
| FQ213239.1 | - | 5 | 590 | |||

Ndufs8
Genomic Map