siRNA Details
| Gene Symbol: Doc2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011937.1 | NP_001011937.1 | 7 | 813 | ||
| XM_006230735.2 | XP_006230797.1 | 7 | 1837 | |||
| GenBank mRNA | BC083779.1 | - | 7 | 813 | 7193879 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Doc2g| Gene Symbol: Doc2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011937.1 | NP_001011937.1 | 7 | 813 | ||
| XM_006230735.2 | XP_006230797.1 | 7 | 1837 | |||
| GenBank mRNA | BC083779.1 | - | 7 | 813 | 7193879 | |