siRNA Details
| Gene Symbol: RGD1307603 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006230736.1 | XP_006230798.1 | 5, 6 | 487 | ||
| XM_006230737.1 | XP_006230799.1 | 5, 6 | 475 | |||
| XM_008760100.1 | XP_008758322.1 | 5, 6 | 546 | |||
| GenBank mRNA | FQ219091.1 | - | 5, 6 | 380 | ||

RGD1307603
Genomic Map