siRNA Details
| Gene Symbol: RGD1307603 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134508.1 | NP_001127980.1 | 2, 3 | 193 | ||
| XM_006230736.1 | XP_006230798.1 | 2, 3 | 188 | |||
| GenBank mRNA | FQ219091.1 | - | 2, 3 | 81 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1307603| Gene Symbol: RGD1307603 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134508.1 | NP_001127980.1 | 2, 3 | 193 | ||
| XM_006230736.1 | XP_006230798.1 | 2, 3 | 188 | |||
| GenBank mRNA | FQ219091.1 | - | 2, 3 | 81 | ||