siRNA Details
| Gene Symbol: Eif1ad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008305.1 | NP_001008306.1 | 3, 4 | 744 | ||
| GenBank mRNA | BC085818.1 | - | 3, 4 | 744 | 7124507 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eif1ad| Gene Symbol: Eif1ad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008305.1 | NP_001008306.1 | 3, 4 | 744 | ||
| GenBank mRNA | BC085818.1 | - | 3, 4 | 744 | 7124507 | |