siRNA Details
| Gene Symbol: Chkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017177.1 | NP_058873.1 | 6 | 992 | ||
| XM_006242181.2 | XP_006242243.2 | 6 | 1658 | |||
| XM_008765677.1 | XP_008763899.1 | 6 | 1655 | |||
| GenBank mRNA | AB006607.1 | - | 6 | 992 | ||
| BC060515.1 | - | 6 | 968 | 5598184 | ||

Chkb
Genomic Map