siRNA Details
Gene Symbol: Ehd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011939.2 | NP_001011939.1 | 2 | 507 | ||
GenBank mRNA | BC082030.1 | - | 2 | 461 | ||
BC160908.1 | - | 2 | 508 | |||
CV120970.1 | - | 2 | 507 |
