siRNA Details
| Gene Symbol: Fgf18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019199.1 | NP_062072.1 | 3 | 560 | ||
| GenBank mRNA | AB004638.1 | - | 3 | 560 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fgf18| Gene Symbol: Fgf18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019199.1 | NP_062072.1 | 3 | 560 | ||
| GenBank mRNA | AB004638.1 | - | 3 | 560 | ||