siRNA Details
| Gene Symbol: Esrra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008511.2 | NP_001008511.2 | 2 | 139 | ||
| XM_006230755.1 | XP_006230817.1 | 2 | 252 | |||
| GenBank mRNA | AY280663.1 | - | 1 | 33 | ||
| BC104701.1 | - | 2 | 139 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Esrra| Gene Symbol: Esrra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008511.2 | NP_001008511.2 | 2 | 139 | ||
| XM_006230755.1 | XP_006230817.1 | 2 | 252 | |||
| GenBank mRNA | AY280663.1 | - | 1 | 33 | ||
| BC104701.1 | - | 2 | 139 | |||