siRNA Details
| Gene Symbol: Rasa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031574.1 | NP_113762.1 | 14, 15 | 1425 | ||
| GenBank mRNA | D86045.1 | - | 14, 15 | 1425 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rasa3| Gene Symbol: Rasa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031574.1 | NP_113762.1 | 14, 15 | 1425 | ||
| GenBank mRNA | D86045.1 | - | 14, 15 | 1425 | ||