siRNA Details
| Gene Symbol: Rasa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031574.1 | NP_113762.1 | 24 | 2489 | ||
| GenBank mRNA | AB028626.1 | - | 346 | |||
| D86045.1 | - | 24 | 2489 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rasa3| Gene Symbol: Rasa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031574.1 | NP_113762.1 | 24 | 2489 | ||
| GenBank mRNA | AB028626.1 | - | 346 | |||
| D86045.1 | - | 24 | 2489 | |||