siRNA Details
| Gene Symbol: Ms4a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106336.1 | NP_001099806.1 | 2, 3 | 261 | ||
| XM_006231032.2 | XP_006231094.1 | 2, 3 | 538 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ms4a10| Gene Symbol: Ms4a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106336.1 | NP_001099806.1 | 2, 3 | 261 | ||
| XM_006231032.2 | XP_006231094.1 | 2, 3 | 538 | |||