siRNA Details
| Gene Symbol: Uncx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017179.1 | NP_058875.1 | 3 | 1619 | ||
| GenBank mRNA | D87748.1 | - | 3 | 1619 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uncx| Gene Symbol: Uncx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017179.1 | NP_058875.1 | 3 | 1619 | ||
| GenBank mRNA | D87748.1 | - | 3 | 1619 | ||