siRNA Details
| Gene Symbol: Psat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198738.2 | NP_942033.2 | 4 | 383 | ||
| GenBank mRNA | AY239016.1 | - | 4 | 371 | ||
| BC079352.1 | - | 4 | 383 | 7108218 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psat1| Gene Symbol: Psat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198738.2 | NP_942033.2 | 4 | 383 | ||
| GenBank mRNA | AY239016.1 | - | 4 | 371 | ||
| BC079352.1 | - | 4 | 383 | 7108218 | ||