siRNA Details
| Gene Symbol: Mcee | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106341.1 | NP_001099811.1 | 2 | 384 | ||
| XM_008759512.1 | XP_008757734.1 | 2 | 332 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mcee| Gene Symbol: Mcee | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106341.1 | NP_001099811.1 | 2 | 384 | ||
| XM_008759512.1 | XP_008757734.1 | 2 | 332 | |||