siRNA Details
| Gene Symbol: Dusp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037973.1 | NP_001033062.1 | 3 | 929 | ||
| GenBank mRNA | BC110044.1 | - | 3 | 929 | 7462125 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dusp9| Gene Symbol: Dusp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037973.1 | NP_001033062.1 | 3 | 929 | ||
| GenBank mRNA | BC110044.1 | - | 3 | 929 | 7462125 | |