siRNA Details
| Gene Symbol: Cdc37l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011941.1 | NP_001011941.1 | 2 | 325 | ||
| XM_006231201.2 | XP_006231263.1 | 3 | 484 | |||
| XM_006231202.2 | XP_006231264.1 | 3 | 484 | |||
| XM_008760322.1 | XP_008758544.1 | 3 | 484 | |||
| GenBank mRNA | BC083761.1 | - | 2 | 325 | 7192975 | |

Cdc37l1
Genomic Map