siRNA Details
| Gene Symbol: Tnni2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017185.1 | NP_058881.1 | 4 | 114 | ||
| GenBank mRNA | M73701.1 | - | 4 | 114 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnni2| Gene Symbol: Tnni2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017185.1 | NP_058881.1 | 4 | 114 | ||
| GenBank mRNA | M73701.1 | - | 4 | 114 | ||