siRNA Details
| Gene Symbol: Fmr1nb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229540.2 | XP_006229602.1 | 4 | 666 | ||
| XM_008773613.1 | XP_008771835.1 | 5 | 684 | |||
| XM_215239.7 | XP_215239.2 | 5 | 684 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fmr1nb| Gene Symbol: Fmr1nb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229540.2 | XP_006229602.1 | 4 | 666 | ||
| XM_008773613.1 | XP_008771835.1 | 5 | 684 | |||
| XM_215239.7 | XP_215239.2 | 5 | 684 | |||