siRNA Details
| Gene Symbol: Col1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053304.1 | NP_445756.1 | 48 | 3741 | ||
| GenBank mRNA | BC133728.1 | - | 48 | 3725 | 7125649 | |
| KJ696743.1 | - | 4 | 337 | |||
| Z78279.1 | - | 48 | 3630 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Col1a1| Gene Symbol: Col1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053304.1 | NP_445756.1 | 48 | 3741 | ||
| GenBank mRNA | BC133728.1 | - | 48 | 3725 | 7125649 | |
| KJ696743.1 | - | 4 | 337 | |||
| Z78279.1 | - | 48 | 3630 | |||