siRNA Details
| Gene Symbol: Wnt8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106359.1 | NP_001099829.1 | 4 | 359 | ||
| XM_006231442.2 | XP_006231504.1 | 5 | 515 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wnt8b| Gene Symbol: Wnt8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106359.1 | NP_001099829.1 | 4 | 359 | ||
| XM_006231442.2 | XP_006231504.1 | 5 | 515 | |||