siRNA Details
Gene Symbol: Wnt8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106359.1 | NP_001099829.1 | 5 | 919 | ||
XM_006231442.2 | XP_006231504.1 | 6 | 1075 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Wnt8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106359.1 | NP_001099829.1 | 5 | 919 | ||
XM_006231442.2 | XP_006231504.1 | 6 | 1075 |