siRNA Details
| Gene Symbol: Papss2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106375.2 | NP_001099845.2 | 10 | 1476 | ||
| XM_006231274.2 | XP_006231336.1 | 11 | 1636 | |||
| GenBank mRNA | BC169061.1 | - | 10 | 1476 | 5621354 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Papss2| Gene Symbol: Papss2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106375.2 | NP_001099845.2 | 10 | 1476 | ||
| XM_006231274.2 | XP_006231336.1 | 11 | 1636 | |||
| GenBank mRNA | BC169061.1 | - | 10 | 1476 | 5621354 | |