siRNA Details
| Gene Symbol: LOC294154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039607.1 | NP_001034696.1 | 2 | 230 | ||
| XM_008772799.1 | XP_008771021.1 | 2 | 424 | |||
| XM_008772800.1 | XP_008771022.1 | 2 | 538 | |||
| XM_008772801.1 | XP_008771023.1 | 2 | 963 | |||
| GenBank mRNA | BC112320.1 | - | 2 | 230 | 7933524 | |

LOC294154
Genomic Map