siRNA Details
Gene Symbol: Olr1730 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000279.1 | NP_001000279.1 | 1 | 738 |
Gene Symbol: Olr1869 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213732.1 | NP_998897.1 | 1 | 738 |
