siRNA Details
| Gene Symbol: Ppp1r11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212542.3 | NP_997707.2 | 1 | 233 | ||
| GenBank mRNA | BC079252.1 | - | 1 | 202 | 7133103 | |
| EX488885.1 | - | 1 | 227 | |||
| FM088354.1 | - | 1 | 232 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp1r11| Gene Symbol: Ppp1r11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212542.3 | NP_997707.2 | 1 | 233 | ||
| GenBank mRNA | BC079252.1 | - | 1 | 202 | 7133103 | |
| EX488885.1 | - | 1 | 227 | |||
| FM088354.1 | - | 1 | 232 | |||