siRNA Details
| Gene Symbol: Ppp1r11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212542.3 | NP_997707.2 | 3 | 419 | ||
| XM_006255870.1 | XP_006255932.1 | 3 | 484 | |||
| GenBank mRNA | BC079252.1 | - | 3 | 388 | 7133103 | |
| EX488885.1 | - | 3 | 413 | |||
| FM088354.1 | - | 3 | 418 | |||

Ppp1r11
Genomic Map